Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-(Bromomethyl)phenyl)acetic acid 96%

Supplier: AMBEED, INC
Danger

A540602-10G A540602-1G A540602-25G A540602-5G
77266-110EA 24 USD InStock
77266-110 77266-112 77266-114 77266-116
(4-(Bromomethyl)phenyl)acetic acid 96%
(4-(Bromomethyl)phenyl)acetic acid
Precaución: for Research Use Only


Formula: (C₆H₄(CH₂Br))CH₂COOH
MDL Number: MFCD00010632
CAS Number: 13737-36-5
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty