Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,4,5-Tetrachloroanisole 95%

Supplier: AMBEED, INC
Warning

A540849-250MG A540849-1G
77253-692EA 21 USD InStock
77253-692 77747-106
2,3,4,5-Tetrachloroanisole 95%
2,3,4,5-Tetrachloroanisole
Precaución: for Research Use Only


Formula: C₇H₄Cl₄O
MDL Number: MFCD00012240
CAS Number: 938-86-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty