Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Indol-2-amine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A544043-100MG A544043-250MG
77266-148EA 150 USD InStock
77266-148 77747-110
1H-Indol-2-amine hydrochloride 98%
1H-Indol-2-amine hydrochloride
Precaución: for Research Use Only


Formula: C₈H₉ClN₂
MDL Number: MFCD06798215
CAS Number: 27878-37-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty