Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

10-(Benzyloxy)-10-oxodecanoic acid 97%

Supplier: AMBEED, INC
Warning

A544329-250MG A544329-5G
77266-150EA 41 USD InStock
77266-150 77266-152
10-(Benzyloxy)-10-oxodecanoic acid 97%
10-(Benzyloxy)-10-oxodecanoic acid
Precaución: for Research Use Only


Formula: C₁₇H₂₄O₄
MW: 292.37 g/mol
MDL Number: MFCD30589211
CAS Number: 67852-88-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty