Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Bromo-5-methylhexane 95%

Supplier: AMBEED, INC
Warning

A544903-100MG A544903-10G A544903-1G A544903-250MG A544903-25G A544903-5G
77241-912EA 12 USD InStock
77241-912 77241-914 77241-916 77241-918 77241-920 77241-922
1-Bromo-5-methylhexane 95%
1-Bromo-5-methylhexane
Precaución: for Research Use Only


Formula: C₇H₁₅Br
MDL Number: MFCD00041674
CAS Number: 35354-37-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty