Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-1-bromocyclobutanecarboxylate 95%

Supplier: AMBEED, INC
Danger

A545437-100MG A545437-10G A545437-1G A545437-250MG A545437-25G A545437-5G
77241-936EA 5 USD InStock
77241-936 77241-938 77241-940 77241-942 77241-944 77241-946
Ethyl-1-bromocyclobutanecarboxylate 95%
Ethyl-1-bromocyclobutanecarboxylate
Precaución: for Research Use Only


Formula: C₇H₁₁BrO₂
MDL Number: MFCD00001322
CAS Number: 35120-18-4
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty