Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Methyl 3-(4-bromophenyl)-2-((tert-butoxycarbonyl)amino)propanoate 98%

Supplier: AMBEED, INC
Warning

A547618-100MG A547618-1G A547618-250MG A547618-5G
77241-948EA 21 USD InStock
77241-948 77241-950 77747-118 77747-119
(S)-Methyl 3-(4-bromophenyl)-2-((tert-butoxycarbonyl)amino)propanoate 98%
(S)-Methyl 3-(4-bromophenyl)-2-((tert-butoxycarbonyl)amino)propanoate
Precaución: for Research Use Only


Formula: C₁₅H₂₀BrNO₄
MDL Number: MFCD14636392
CAS Number: 266306-18-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty