Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-((tert-Butyldimethylsilyl)oxy)pyrrolidine 97%

Supplier: AMBEED, INC
Danger

A547634-100MG A547634-1G A547634-250MG
77241-952EA 43 USD InStock
77241-952 77241-954 77241-956
(S)-3-((tert-Butyldimethylsilyl)oxy)pyrrolidine 97%
(S)-3-((tert-Butyldimethylsilyl)oxy)pyrrolidine
Precaución: for Research Use Only


Formula: C₁₀H₂₃NOSi
MW: 201.38 g/mol
MDL Number: MFCD09909497
CAS Number: 207113-36-8
UN: 1760
ADR: 8 Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty