Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-3-(trifluoromethyl)benzyl bromide 98%

Supplier: AMBEED, INC
Danger

A547716-1G A547716-250MG A547716-5G
77266-182EA 34 USD InStock
77266-182 77266-184 77266-186
2-Chloro-3-(trifluoromethyl)benzyl bromide 98%
2-Chloro-3-(trifluoromethyl)benzyl bromide
Precaución: for Research Use Only


Formula: C₈H₅BrClF₃
MW: 273.48
MDL Number: MFCD01631539
CAS Number: 261763-22-8
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty