Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-4-Isopropyl-2-(pyridin-2-yl)-4,5-dihydrooxazole ≥98%, ee 99%

Supplier: AMBEED, INC
Warning

A552666-100MG A552666-1G A552666-250MG A552666-25G A552666-5G
77241-982EA 10 USD InStock
77241-982 77241-984 77241-986 77241-988 77241-990
(S)-4-Isopropyl-2-(pyridin-2-yl)-4,5-dihydrooxazole ≥98%, ee 99%
(S)-4-Isopropyl-2-(pyridin-2-yl)-4,5-dihydrooxazole
Precaución: for Research Use Only


Formula: C₁₁H₁₄N₂O
MDL Number: MFCD28016024
CAS Number: 108915-04-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty