Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl 2,2'-Bipyridine-6,6'-dicarboxylate 97%

Supplier: AMBEED, INC
Warning

A553823-100MG A553823-1G A553823-250MG A553823-5G
77253-870EA 8 USD InStock
77253-870 77253-872 77253-874 77253-876
Dimethyl 2,2'-Bipyridine-6,6'-dicarboxylate 97%
Dimethyl 2,2'-Bipyridine-6,6'-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₄H₁₂N₂O₄
MDL Number: MFCD11042478
CAS Number: 142593-07-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty