Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-1-[(R)-1-(Di-tert-butylphosphino)ethyl]-2-[(R)-phenylphosphinoyl]ferrocene 98%

Supplier: AMBEED, INC
Warning

A554271-100MG A554271-250MG A554271-50MG
77253-878EA 103 USD InStock
77253-878 77253-880 77253-882
(R)-1-[(R)-1-(Di-tert-butylphosphino)ethyl]-2-[(R)-phenylphosphinoyl]ferrocene 98%
(R)-1-[(R)-1-(Di-tert-butylphosphino)ethyl]-2-[(R)-phenylphosphinoyl]ferrocene
Precaución: for Research Use Only


Formula: C₂₆H₃₆FeOP₂
MDL Number: MFCD18827472
CAS Number: 1221746-56-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty