Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl (2-((R)-(3-chlorophenyl)((R)-1-(((S)-2-(methylamino)-3-((R)-tetrahydro-2H-pyran-3-yl)propyl)carbamoyl)piperidin-3-yl)methoxy)ethyl)carbamate 2,2,2-trifluoroacetate 98+%

Supplier: AMBEED, INC
Warning

A556461-10MG A556461-1MG A556461-5MG
77241-992EA 150 USD InStock
77241-992 77241-994 77241-996
Methyl (2-((R)-(3-chlorophenyl)((R)-1-(((S)-2-(methylamino)-3-((R)-tetrahydro-2H-pyran-3-yl)propyl)carbamoyl)piperidin-3-yl)methoxy)ethyl)carbamate 2,2,2-trifluoroacetate 98+%
Methyl (2-((R)-(3-chlorophenyl)((R)-1-(((S)-2-(methylamino)-3-((R)-tetrahydro-2H-pyran-3-yl)propyl)carbamoyl)piperidin-3-yl)methoxy)ethyl)carbamate 2,2,2-trifluoroacetate
Precaución: for Research Use Only


Formula: C₂₈H₄₂ClF₃N₄O₇
MW: 639.1 g/mol
MDL Number: MFCD20278230
CAS Number: 1013937-63-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty