Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Salermide 98%

Supplier: AMBEED, INC
Warning

A557147-10MG A557147-1MG A557147-25MG A557147-50MG A557147-5MG
77266-296EA 11 USD InStock
77266-296 77266-298 77266-300 77266-302 77266-304
Salermide 98%
Salermide
Precaución: for Research Use Only


Formula: C₂₆H₂₂N₂O₂
MDL Number: MFCD12912446
CAS Number: 1105698-15-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty