Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-methyl-4-pentenoate 96%

Supplier: AMBEED, INC
Danger

A560973-100MG A560973-1G A560973-250MG
77242-056EA 9 USD InStock
77242-056 77242-058 77242-060
Ethyl-4-methyl-4-pentenoate 96%
Ethyl-4-methyl-4-pentenoate
Precaución: For research use only.


Formula: C₈H₁₄O₂
MW: 142.198 g/mol
MDL Number: MFCD00075160
CAS Number: 4911-54-0
UN: 3272
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty