Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Isopropylnicotinic acid hydrochloride 98%

Supplier: AMBEED, INC
Warning

A561770-100MG A561770-250MG A561770-1G
77266-350EA 124 USD InStock
77266-350 77266-352 77749-160
6-Isopropylnicotinic acid hydrochloride 98%
6-Isopropylnicotinic acid hydrochloride
Precaución: for Research Use Only


Formula: C₉H₁₂ClNO₂
MDL Number: MFCD22421844
CAS Number: 1423029-80-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty