Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chlorobenzenesulfonyl chloride 98% (by GC)

Supplier: AMBEED, INC
Danger

A561876-10G A561876-1G A561876-25G A561876-5G
77280-048EA 22 USD InStock
77280-048 77280-050 77280-052 77280-054
3-Chlorobenzenesulfonyl chloride 98% (by GC)
3-Chlorobenzenesulphonyl chloride
Precaución: for Research Use Only


Formula: C₆H₄Cl₂O₂S
MDL Number: MFCD00051697
CAS Number: 2888-06-4
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty