Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3,5-difluoroaniline 97%

Supplier: AMBEED, INC
Danger

A562466-1G A562466-25G A562466-5G
77292-544EA 18 USD InStock
77292-544 77292-546 77292-548
4-Bromo-3,5-difluoroaniline 97%
4-Bromo-3,5-difluoroaniline
Precaución: for Research Use Only


Formula: C₆H₄BrF₂N
MDL Number: MFCD00236593
CAS Number: 203302-95-8
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty