Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ME-0328 98%

Supplier: AMBEED, INC
Warning

A562763-10MG A562763-1MG A562763-25MG A562763-50MG A562763-5MG
77266-354EA 47 USD InStock
77266-354 77266-356 77266-358 77266-360 77266-362
ME-0328 98%
ME-0328
Precaución: for Research Use Only


Formula: C₁₉H₁₉N₃O₂
MDL Number: MFCD27991291
CAS Number: 1445251-22-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty