Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Acetylphenyl trifluoromethanesulfonate 95%

Supplier: AMBEED, INC
Danger

A564421-1G A564421-250MG A564421-25G A564421-5G
77266-386EA 12 USD InStock
77266-386 77266-388 77266-390 77266-392
4-Acetylphenyl trifluoromethanesulfonate 95%
4-Acetylphenyl trifluoromethanesulfonate
Precaución: for Research Use Only


Formula: C₉H₇F₃O₄S
MDL Number: MFCD00191706
CAS Number: 109613-00-5
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty