Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Dodecylphenol, mixture of isomers 95%

Supplier: AMBEED, INC
Danger

A564682-10G A564682-25G
77292-562EA 6 USD InStock
77292-562 77307-820
4-Dodecylphenol, mixture of isomers 95%
4-Dodecylphenol, mixture of isomers
Precaución: for Research Use Only


Formula: C₁₂H₂₅C₆H₄OH
MDL Number: MFCD00039521
CAS Number: 27193-86-8
UN: 3145
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty