Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-3-(hydroxymethyl)benzaldehyde 97%

Supplier: AMBEED, INC
Warning

A564722-100MG A564722-1G A564722-250MG
77242-156EA 34 USD InStock
77242-156 77242-158 77242-160
4-Bromo-3-(hydroxymethyl)benzaldehyde 97%
4-Bromo-3-(hydroxymethyl)benzaldehyde
Precaución: for Research Use Only


Formula: C₈H₇BrO₂
MW: 215.04 g/mol
MDL Number: MFCD29762439
CAS Number: 254744-15-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty