Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

M2I-1 ≥98%

Supplier: AMBEED, INC
Warning

A566234-10MG A566234-1MG A566234-25MG A566234-50MG A566234-5MG
77242-200EA 82 USD InStock
77242-200 77242-202 77242-204 77242-206 77242-208
M2I-1 ≥98%
M2I-1
Precaución: for Research Use Only


Formula: C₁₉H₂₄N₄O₄S
MDL Number: MFCD00469459
CAS Number: 312271-03-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty