Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Diaminobenzenesulfonamide 95%

Supplier: AMBEED, INC
Warning

A567013-100MG A567013-1G A567013-250MG
77280-074EA 112 USD InStock
77280-074 77280-076 77280-078
3,4-Diaminobenzenesulfonamide 95%
3,4-Diaminobenzenesulfonamide
Precaución: for Research Use Only


Formula: C₆H₉N₃O₂S
MDL Number: MFCD00277940
CAS Number: 2360-20-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty