Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

[2,2':6',2''-Terpyridine]-4'-carbaldehyde 97%

Supplier: AMBEED, INC
Warning

A568863-100MG A568863-250MG A568863-5G
77254-028EA 25 USD InStock
77254-028 77254-030 77747-171
[2,2':6',2''-Terpyridine]-4'-carbaldehyde 97%
[2,2':6',2''-Terpyridine]-4'-carbaldehyde
Precaución: for Research Use Only


Formula: C₁₆H₁₁N₃O
MW: 261.27 g/mol
MDL Number: MFCD06796962
CAS Number: 108295-45-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty