Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CZ415 98+%

Supplier: AMBEED, INC
Warning

A569091-10MG A569091-1MG A569091-25MG A569091-50MG A569091-5MG
77266-418EA 50 USD InStock
77266-418 77266-420 77266-422 77266-424 77266-426
CZ415 98+%
CZ415
Precaución: for Research Use Only


Formula: C₂₂H₂₉N₅O₄S
MDL Number: MFCD30533321
CAS Number: 1429639-50-8
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty