Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Benzyloxyaniline 95%

Supplier: AMBEED, INC
Warning

A569612-100G A569612-10G A569612-1G A569612-25G A569612-5G
77266-428EA 264 USD InStock
77266-428 77266-430 77266-432 77266-434 77266-436
2-Benzyloxyaniline 95%
2-Benzyloxyaniline
Precaución: for Research Use Only


Formula: C₁₃H₁₃NO
MW: 199.25
MDL Number: MFCD03093873
CAS Number: 20012-63-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty