Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

CD437 98%

Supplier: AMBEED, INC
Danger

A570556-10MG A570556-1MG A570556-25MG A570556-5MG
77254-050EA 68 USD InStock
77254-050 77254-052 77254-054 77254-056
CD437 98%
CD437
Precaución: for Research Use Only


Formula: C₂₇H₂₆O₃
MDL Number: MFCD03106506
CAS Number: 125316-60-1
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty