Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-4-hydroxycoumarin 98%

Supplier: AMBEED, INC
Warning

A571771-1G A571771-250MG A571771-5G A571771-25G
77266-446EA 25 USD InStock
77266-446 77266-448 77266-450 77747-183
6-Chloro-4-hydroxycoumarin 98%
6-Chloro-4-hydroxycoumarin
Precaución: for Research Use Only


Formula: C₉H₅ClO₃
MW: 196.59 g/mol
MDL Number: MFCD00239399
CAS Number: 19484-57-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty