Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-bromo-3-methoxybenzonitrile 97%

Supplier: AMBEED, INC
Danger

A571783-1G A571783-250MG A571783-25G A571783-5G
77266-452EA 23 USD InStock
77266-452 77266-454 77266-456 77266-458
2-Amino-5-bromo-3-methoxybenzonitrile 97%
2-Amino-5-bromo-3-methoxybenzonitrile
Precaución: for Research Use Only


Formula: C₈H₇BrN₂O
MDL Number: MFCD22570412
CAS Number: 176718-54-0
UN: 3439
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty