Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

VO-OHpic trihydrate 98%

Supplier: AMBEED, INC
Warning

A573257-100MG A573257-10MG A573257-1MG A573257-50MG A573257-5MG
77242-270EA 339 USD InStock
77242-270 77242-272 77242-274 77242-276 77242-278
VO-OHpic trihydrate 98%
VO-OHpic trihydrate
Precaución: for Research Use Only


Formula: C₁₂H₁₁N₂O₉V
MW: 378.1653 g/mol
MDL Number: MFCD11045907
CAS Number: 476310-60-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty