Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Methylveratrole 95%

Supplier: AMBEED, INC
Warning

A573655-10G A573655-1G A573655-5G A573655-25G A573655-100G
77254-076EA 66 USD InStock
77254-076 77254-078 77254-080 77747-187 77747-186
3-Methylveratrole 95%
3-Methylveratrole
Precaución: for Research Use Only


Formula: C₉H₁₂O₂
MDL Number: MFCD00008379
CAS Number: 4463-33-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty