Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Di(pyridin-4-yl)-1,3,4-thiadiazole 95%

Supplier: AMBEED, INC
Warning

A573726-100MG A573726-250MG A573726-5G
77254-082EA 33 USD InStock
77254-082 77254-084 77254-086
2,5-Di(pyridin-4-yl)-1,3,4-thiadiazole 95%
2,5-Di(pyridin-4-yl)-1,3,4-thiadiazole
Precaución: for Research Use Only


Formula: C₁₂H₈N₄S
MDL Number: MFCD00044193
CAS Number: 15311-09-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty