Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4aR,6aS,6bR,8aS,12aS,12bR,14bS)-8a-(1H-Imidazole-1-carbonyl)-4,4,6a,6b,11,11,14b-heptamethyl-3,13-dioxo-3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-octadecahydropicene-2-carbonitrile 98%

Supplier: AMBEED, INC
Warning

A574651-10MG A574651-1MG A574651-5MG
77242-294EA 57 USD InStock
77242-294 77242-296 77242-298
(4aR,6aS,6bR,8aS,12aS,12bR,14bS)-8a-(1H-Imidazole-1-carbonyl)-4,4,6a,6b,11,11,14b-heptamethyl-3,13-dioxo-3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-octadecahydropicene-2-carbonitrile 98%
(4aR,6aS,6bR,8aS,12aS,12bR,14bS)-8a-(1H-Imidazole-1-carbonyl)-4,4,6a,6b,11,11,14b-heptamethyl-3,13-dioxo-3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-octadecahydropicene-2-carbonitrile
Precaución: for Research Use Only


Formula: C₃₄H₄₃N₃O₃
MDL Number: MFCD12756341
CAS Number: 443104-02-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty