Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

p-Toluenesulfonylacetonitrile 97%

Supplier: AMBEED, INC
Danger

A574786-10G A574786-1G A574786-25G A574786-5G
77280-134EA 11 USD InStock
77280-134 77280-136 77280-138 77280-140
p-Toluenesulfonylacetonitrile 97%
p-Toluenesulphonylacetonitrile
Precaución: for Research Use Only


Formula: C₉H₉NO₂S
MDL Number: MFCD00039487
CAS Number: 5697-44-9
UN: 3439
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty