Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromophenyl methyl sulphone 97%

Supplier: AMBEED, INC
Warning

A576853-10G A576853-1G A576853-250MG A576853-25G A576853-5G
77242-326EA 76 USD InStock
77242-326 77242-328 77242-330 77242-332 77242-334
2-Bromophenyl methyl sulphone 97%
2-Bromophenyl methyl sulphone
Precaución: for Research Use Only


Formula: C₇H₇BrO₂S
MW: 235.1
MDL Number: MFCD07368924
CAS Number: 33951-33-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty