Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(sec-Butyl)-4,5-dihydrothiazole 97%

Supplier: AMBEED, INC
Warning

A578012-100MG A578012-250MG A578012-5G
77242-368EA 23 USD InStock
77242-368 77242-370 77242-372
2-(sec-Butyl)-4,5-dihydrothiazole 97%
2-(sec-Butyl)-4,5-dihydrothiazole
Precaución: for Research Use Only


Formula: C₇H₁₃NS
MDL Number: MFCD28044285
CAS Number: 56367-27-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty