Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

KS-176 98%

Supplier: AMBEED, INC
Warning

A578694-100MG A578694-10MG A578694-1MG A578694-25MG A578694-50MG A578694-5MG
77242-394EA 143 USD InStock
77242-394 77242-396 77242-398 77242-400 77242-402 77242-404
KS-176 98%
KS-176
Precaución: for Research Use Only


Formula: C₂₂H₁₉N₃O₅
MDL Number: MFCD22666402
CAS Number: 1253452-78-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty