Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3,5,7-Tetrakis(4-iodophenyl)adamantane 97%

Supplier: AMBEED, INC
Warning

A579769-1G A579769-250MG
77242-410EA 80 USD InStock
77242-410 77242-412
1,3,5,7-Tetrakis(4-iodophenyl)adamantane 97%
1,3,5,7-Tetrakis(4-iodophenyl)adamantane
Precaución: for Research Use Only


Formula: C₃₄H₂₈I₄
MW: 944.20 g/mol
MDL Number: MFCD30187648
CAS Number: 144970-30-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty