Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(Triisopropoxysilyl)propyl methacrylate 97%

Supplier: AMBEED, INC
Warning

A581656-1G A581656-25G A581656-5G
77254-146EA 9 USD InStock
77254-146 77254-148 77747-206
3-(Triisopropoxysilyl)propyl methacrylate 97%
3-(Triisopropoxysilyl)propyl methacrylate
Precaución: for Research Use Only


Formula: C₁₆H₃₂O₅Si
MDL Number: MFCD08275516
CAS Number: 80750-05-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty