Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloro(pyridine)bis(dimethylglyoximato)cobalt(III) 98%

Supplier: AMBEED, INC
Warning

A582236-5G A582236-1G
77266-542EA 133 USD InStock
77266-542 77551-748
Chloro(pyridine)bis(dimethylglyoximato)cobalt(III) 98%
Chloro(pyridine)bis(dimethylglyoximato)cobalt(III)
Precaución: for Research Use Only


Formula: C₁₃H₁₉ClCoN₅O₄
MDL Number: MFCD00075236
CAS Number: 23295-32-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty