Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Fluoroorotic acid 96%

Supplier: AMBEED, INC
Warning

A587031-1G A587031-25G A587031-5G A587031-10G
77292-668EA 27 USD InStock
77292-668 77292-670 77617-466 77749-206
5-Fluoroorotic acid 96%
5-Fluoroorotic acid
Precaución: for Research Use Only


Formula: C₅H₃FN₂O₄
MW: 174.09 g/mol
MDL Number: MFCD00042526
CAS Number: 703-95-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty