Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Ethylnaphthalene 99% (by GC)

Supplier: AMBEED, INC
Danger

A587804-1G A587804-25G A587804-5G
77266-568EA 16 USD InStock
77266-568 77266-570 77266-572
2-Ethylnaphthalene 99% (by GC)
2-Ethylnaphthalene
Precaución: for Research Use Only


Formula: C₁₂H₁₂
MDL Number: MFCD00004127
CAS Number: 939-27-5
UN: 3082
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty