Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Anilinopyridine 95%

Supplier: AMBEED, INC
Warning

A588359-1G A588359-250MG A588359-5G
77292-694EA 57 USD InStock
77292-694 77292-696 77292-698
4-Anilinopyridine 95%
4-Anilinopyridine
Precaución: for Research Use Only


Formula: C₁₁H₁₀N₂
MDL Number: MFCD00457758
CAS Number: 22961-45-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty