Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ML390 98+%

Supplier: AMBEED, INC
Warning

A589360-10MG A589360-1MG A589360-25MG A589360-5MG
77266-596EA 69 USD InStock
77266-596 77266-598 77266-600 77266-602
ML390 98+%
ML390
Precaución: for Research Use Only


Formula: C₂₁H₂₁F₃N₂O₃
MDL Number: MFCD30489750
CAS Number: 2029049-79-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty