Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromo-6-methyl-1H-indazole 98%

Supplier: AMBEED, INC
Warning

A589501-100MG A589501-1G A589501-250MG
77292-700EA 90 USD InStock
77292-700 77292-702 77292-704
4-Bromo-6-methyl-1H-indazole 98%
4-Bromo-6-methyl-1H-indazole
Precaución: for Research Use Only


Formula: C₈H₇BrN₂
MDL Number: MFCD07781508
CAS Number: 885521-94-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty