Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DMH-1 ≥99%

Supplier: AMBEED, INC
Warning

A590459-1MG A590459-25MG A590459-50MG A590459-5MG
77242-510EA 36 USD InStock
77242-510 77242-512 77242-514 77242-516
DMH-1 ≥99%
DMH-1
Precaución: for Research Use Only


Formula: C₂₄H₂₀N₄O
MDL Number: MFCD18384963
CAS Number: 1206711-16-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty