Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-1-(1-pyrrolidinyl)-1-ethanone hydrochloride 95%

Supplier: AMBEED, INC
Warning

A594040-1G A594040-250MG
77254-204EA 41 USD InStock
77254-204 77254-206
2-Amino-1-(1-pyrrolidinyl)-1-ethanone hydrochloride 95%
2-Amino-1-(1-pyrrolidinyl)-1-ethanone hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₃ClN₂O
MW: 164.64 g/mol
MDL Number: MFCD07366742
CAS Number: 35855-14-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty