Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-hydroxy-5-nitropyridine 96%

Supplier: AMBEED, INC
Warning

A599634-10G A599634-1G A599634-250MG A599634-25G A599634-5G
77280-230EA 17 USD InStock
77280-230 77280-232 77280-234 77280-236 77280-238
3-Chloro-2-hydroxy-5-nitropyridine 96%
3-Chloro-2-hydroxy-5-nitropyridine
Precaución: for Research Use Only


Formula: C5H3ClN2O3
MDL Number: MFCD17015099
CAS Number: 22353-38-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty