Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Indol-6-ylmethylamine 96%

Supplier: AMBEED, INC
Danger

A600582-100MG A600582-250MG A600582-1G
77242-576EA 15 USD InStock
77242-576 77242-578 77569-146
1H-Indol-6-ylmethylamine 96%
1H-Indol-6-ylmethylamine
Precaución: For research use only.


Formula: C₉H₁₀N₂
MW: 146.19 g/mol
MDL Number: MFCD06213839
CAS Number: 3468-17-5
UN: 1759
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty